Learn More
Invitrogen™ NAMPT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534858
Description
Recommended positive controls: 293T, A431, Jurkat, Raji, K562, NCI-H929. Predicted reactivity: Mouse (96%), Rat (96%), Xenopus laevis (92%), Pig (97%), Chicken (96%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Visfatin, also known as pre-B cell colony-enhancing factor, is an adipocytokine that is abundantly secreted by visceral adipose tissue, with insulin mimetic effects. Visfatin mimics insulin signaling by binding to the insulin receptor and reducing glucose release from liver cells, while stimulating glucose utilization in adipocytes and myocytes. Visfatin is also invoved in enhancement of cell proliferation, biosynthesis of nicotinamide mono-and dinucleotide and glucose homeostasis.
Specifications
| NAMPT | |
| Polyclonal | |
| Unconjugated | |
| NAMPT | |
| 1110035O14Rik; AI314458; AI480535; DKFZp666B131; EC 2.4.2.12; hypothetical protein; MGC117256; NAmPRTase; NAMPT; nampt1; nampta; NAMPTP1; Nicotinamide phosphoribosyltransferase; nicotinamide phosphoribosyltransferase 1; nicotinamide phosphoribosyltransferase a; nicotinamide phosphoribosyltransferase pseudogene 1; PBEF; Pbef1; pre-B cell-enhancing factor; pre-B-cell colony enhancing factor 1; pre-B-cell colony-enhancing factor 1; Pre-B-cell colony-enhancing factor 1 homolog; pre-B-cell colony-enhancing factor 1 precursor; VF; VISFATIN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10135, 297508 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P43490, Q80Z29 | |
| NAMPT | |
| Recombinant protein encompassing a sequence within the center region of human Visfatin. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.