missing translation for 'onlineSavingsMsg'
Learn More

Namodenoson, MedChemExpress

Product Code. 16453296
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453296

Brand: Medchem Express HY12365

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Namodenoson (CF-102) is a selective A3 adenosine receptor (A3AR) agonist (Ki=0.33 nM). Namodenoson displays 2500- and 1400-fold selectivity over A1 and A2A receptors respectively.

Namodenoson (CF-102) is a selective A3 adenosine receptor (A3AR) agonist (Ki=0.33 nM). Namodenoson displays 2500- and 1400-fold selectivity over A1 and A2A receptors respectively.

Specifications

Chemical Name or Material Namodenoson
CAS 163042-96-4
Color White
Purity Grade Notes Research
Molecular Formula C18H18ClIN6O4
Quantity 25 mg
Synonym CF-102 2-Cl-IB-MECA
Solubility Information DMSO : ≥ 31 mg/mL (56.91 mM)
SMILES O[C@H]([C@@H]1O)[C@@H](O[C@@H]1C(NC)=O)N2C3=NC(Cl)=NC(NCC4=CC(I)=CC=C4)=C3N=C2
Molecular Weight (g/mol) 544.73
Formula Weight 544.73
Percent Purity 99.6%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.