missing translation for 'onlineSavingsMsg'
Learn More

Naltrexone Hydrochloride, TRC

Product Code. 17910405
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17910405 1 g 1g
17974495 10 g 10g
17962195 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17910405 Supplier Toronto Research Chemicals Supplier No. TRCN2857501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16676-29-2
Molecular Formula C20 H23 N O4 . Cl H
Molecular Weight (g/mol) 377.86
Synonym (5α)-17-(Cyclopropylmethyl)-4,5-epoxy-3,14-dihydroxymorphinan-6-one Hydrochloride (1:1), Morphinan-6-one, 17-(cyclopropylmethyl)-4,5-epoxy-3,14-dihydroxy-, hydrochloride, (5α)- (9CI), Morphinan-6-one, 17-(cyclopropylmethyl)-4,5α-epoxy-3,14-dihydroxy-, hydrochloride (8CI), Antaxone, Depade, EN 1639A, NIH 8503, Nalorex, Naltrexone hydrochloride, ReVia, Trexan
IUPAC Name (4R,4aS,7aR,12bS)-3-(cyclopropylmethyl)-4a,9-dihydroxy-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochloride
SMILES Cl.Oc1ccc2C[C@H]3N(CC4CC4)CC[C@@]56[C@@H](Oc1c25)C(=O)CC[C@@]36O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H23NO4.ClH/c22-13-4-3-12-9-15-20(24)6-5-14(23)18-19(20,16(12)17(13)25-18)7-8-21(15)10-11-1-2-11;/h3-4,11,15,18,22,24H,1-2,5-10H2;1H/t15-,18+,19+,20-;/m1./s1
Formula Weight 377.1394
Percent Purity >95%
Chemical Name or Material Naltrexone hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.