missing translation for 'onlineSavingsMsg'
Learn More

Naltrexone hydrochloride, Thermo Scientific Chemicals

Product Code. 15463339
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15463339 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15463339 Supplier Thermo Scientific Alfa Aesar Supplier No. J60590.MD
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A congener of naloxone and also an opioid antagonist

A congener of naloxone and also an opioid antagonist.Naltrexone hydrochloride is used in the treatment of alcohol and utilized for deceasing drinking habit. It is also used to prevent euphorigenic effects in the treatment of patients addicted to opioids. Further, it is used as a specific opiate antagonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A congener of naloxone and also an opioid antagonist.Naltrexone hydrochloride is used in the treatment of alcohol and utilized for deceasing drinking habit. It is also used to prevent euphorigenic effects in the treatment of patients addicted to opioids. Further, it is used as a specific opiate antagonist.

Solubility
Soluble in water at 50mg/ml.

Notes
Light sensitive. Incompatible with bases and oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16676-29-2
Molecular Formula C20H24ClNO4
Molecular Weight (g/mol) 377.865
MDL Number MFCD00069324
InChI Key RHBRMCOKKKZVRY-ITLPAZOVSA-N
Synonym naltrexone hydrochloride, naltrexone hcl, trexan, depade, antaxone, revia, nalorex, unii-z6375yw9sf, nemexin
PubChem CID 5485201
IUPAC Name (4R,4aS,7aR,12bS)-3-(cyclopropylmethyl)-4a,9-dihydroxy-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochloride
SMILES C1CC1CN2CCC34C5C(=O)CCC3(C2CC6=C4C(=C(C=C6)O)O5)O.Cl

Specifications

Color White
Quantity 250 mg
Beilstein 3580333
Sensitivity Light sensitive
Merck Index 14,6363
Solubility Information Soluble in water at 50mg/ml.
Formula Weight 377.86
Physical Form Powder
Chemical Name or Material Naltrexone hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.