missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number 15480327
Click to view available options
Quantity:
50 mg
100 mg
250 mg
This item is not returnable. View return policy
4238-41-9
C14H22Cl2N2O3S
369.30
MFCD00065395
YFCUZWYIPBUQBD-ZOWNYOTGSA-N
1-chloro-3-tosylamido-7-amino-2-hepta none hydrochloride, p-toluenesulfonamide, hydrochloride 7ci, 8ci, nalpha-p-toluenesulfonyl-dl-lysine chloromethyl ketone hydrochloride
23335676
N-(7-amino-1-chloro-2-oxoheptan-3-yl)-4-methylbenzenesulfonamide;hydrochloride
Cl.CC1=CC=C(C=C1)S(=O)(=O)N[C@@H](CCCCN)C(=O)CCl
This item is not returnable. View return policy
4238-41-9
C14H22Cl2N2O3S
369.30
MFCD00065395
YFCUZWYIPBUQBD-ZOWNYOTGSA-N
1-chloro-3-tosylamido-7-amino-2-hepta none hydrochloride, p-toluenesulfonamide, hydrochloride 7ci, 8ci, nalpha-p-toluenesulfonyl-dl-lysine chloromethyl ketone hydrochloride
23335676
N-(7-amino-1-chloro-2-oxoheptan-3-yl)-4-methylbenzenesulfonamide;hydrochloride
Cl.CC1=CC=C(C=C1)S(=O)(=O)N[C@@H](CCCCN)C(=O)CCl

Inhibitor of papain and trypsin

Inhibitor of papain and trypsin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Solubility Information Soluble in water
Formula Weight 369.31
Percent Purity 98%
Quantity 50 mg
Physical Form Powder
Chemical Name or Material Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'