missing translation for 'onlineSavingsMsg'
Learn More

Nalpha,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt, 98%

Product Code. 15497836
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15497836

missing translation for 'mfr': Thermo Scientific Alfa Aesar H62785.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15098-69-8 | C28H53N3O6 | 527.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15098-69-8
Molecular Formula C28H53N3O6
Molecular Weight (g/mol) 527.75
MDL Number MFCD00038892
InChI Key HRLHJTYAMCGERD-VAMKTSTMNA-N
Synonym boc-lys boc-oh.dcha, boc-lys boc-oh dcha, dicyclohexylamine s-2,6-bis tert-butoxycarbonyl amino hexanoate, n,n'-di-boc-l-lysine dicyclohexylamine salt, boc-lys boc-oh dicyclohexammonium salt, n,n'-bis tert-butoxycarbonyl-l-lysine dicyclohexylamine 1:1, 2s-2,6-bis tert-butoxycarbonyl amino hexanoic acid; dicha, boc-lys boc-ohcha, boc-lys boc-oh?cha, boc-lys boc-ohdcha
PubChem CID 12017189
IUPAC Name (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid;N-cyclohexylcyclohexanamine
SMILES C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 139°C to 143°C
Color White
Beilstein 6262378
Quantity 25 g
Formula Weight 527.74
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Nalpha,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.