missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha,Nepsilon-Di-Boc-L-lysine, 98%
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
Nalpha, Nepsilon-Di-Boc-L-lysine is used as fine chemical intermediate, synthesis intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nalpha, Nepsilon-Di-Boc-L-lysine is used as fine chemical intermediate, synthesis intermediates.
Solubility
Soluble in chloroform, ether, ethyl acetate and methanol
Notes
Keep container tightly closed. Store away from oxidizing agents.
Nalpha, Nepsilon-Di-Boc-L-lysine is used as fine chemical intermediate, synthesis intermediates.
Solubility
Soluble in chloroform, ether, ethyl acetate and methanol
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
| Color | White |
| Solubility Information | Soluble in chloroform,ether,ethyl acetate and methanol |
| Quantity | 100 g |
| Formula Weight | 346.42 |
| Percent Purity | 98% |
| Physical Form | Low Melting Solid |
| Chemical Name or Material | Nalpha,Nepsilon-Di-Boc-L-lysine |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction