missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-S-trityl-D-cysteine, 98%

Product Code. 15418838
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418838 1 g 1g
15428838 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418838 Supplier Thermo Scientific Alfa Aesar Supplier No. H62301.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 167015-11-4 | C37H31NO4S | 585.718 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167015-11-4
Molecular Formula C37H31NO4S
Molecular Weight (g/mol) 585.718
MDL Number MFCD00151922
InChI Key KLBPUVPNPAJWHZ-UUWRZZSWSA-N
Synonym fmoc-d-cys trt-oh, n-fmoc-s-trityl-d-cysteine, fmoc-s-trityl-d-cysteine, n-alpha-9-fluorenylmethyloxycarbonyl-s-trityl-d-cysteine, n-9h-fluoren-9-ylmethoxy carbonyl-s-triphenylmethyl-d-cysteine, d-cysteine, n-9h-fluoren-9-ylmethoxy carbonyl-s-triphenylmethyl, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tritylthio propanoic acid, ambotzfaa1035, pubchem19012
PubChem CID 7168037
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-tritylsulfanylpropanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46

Specifications

Melting Point 172°C to 176°C
Quantity 1 g
Beilstein 7558914
Formula Weight 587.71
Percent Purity 98%
Chemical Name or Material Nalpha-Fmoc-S-trityl-D-cysteine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.