missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine, 95%, Thermo Scientific™

Catalog Number 15438576
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
98930-01-9
C31H36N4O7S
608.71
MFCD00043098,MFCD00065616
LKGHIEITYHYVED-UHFFFAOYNA-N
fmoc-arg mtr-oh, fmoc-l-arg mtr, fmoc-arg mtr, fmoc-l-arg mtr-oh, ambotzfaa1700, fmoc-arginine mtr-oh, fmoc-l-arginine mtr, pubchem10000, fmoc-ng-mtr-l-arginine hemiisopropyl ether, fmoc-arg mtr-oh tlc
3010428
COC1=C(C)C(C)=C(C(C)=C1)S(=O)(=O)NC(N)=NCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
This item is not returnable. View return policy
98930-01-9
C31H36N4O7S
608.71
MFCD00043098,MFCD00065616
LKGHIEITYHYVED-UHFFFAOYNA-N
fmoc-arg mtr-oh, fmoc-l-arg mtr, fmoc-arg mtr, fmoc-l-arg mtr-oh, ambotzfaa1700, fmoc-arginine mtr-oh, fmoc-l-arginine mtr, pubchem10000, fmoc-ng-mtr-l-arginine hemiisopropyl ether, fmoc-arg mtr-oh tlc
3010428
COC1=C(C)C(C)=C(C(C)=C1)S(=O)(=O)NC(N)=NCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
IUPAC Name 5-{[amino(4-methoxy-2,3,6-trimethylbenzenesulfonamido)methylidene]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
Formula Weight 608.71
Percent Purity 95%
Quantity 5 g
Sensitivity Moisture sensitive
Physical Form Powder
Chemical Name or Material Nalpha-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'