missing translation for 'onlineSavingsMsg'
Learn More

NAlpha-Fmoc-NEpsilon-biotinyl-L-lysine, TRC

Product Code. 30400821
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400821 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400821 Supplier Toronto Research Chemicals Supplier No. TRCF6227601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nalpha-Fmoc-Nepsilon-biotinyl-L-lysine
CAS 146987-10-2
Molecular Formula C31H38N4O6S
Quantity 1 g
Synonym Fmoc-Lys(Biotin)-OH, N-alpha-(9-Fluorenylmethyloxycarbonyl)-N-epsilon-biotinyl-L-lysine, (S)-2-(((9H-Fluoren-9-yl)methoxy)carbonylamino)-6-(5-((3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexanoic Acid
InChI Formula InChI=1S/C31H38N4O6S/c36-27(15-6-5-14-26-28-25(18-42-26)33-30(39)35-28)32-16-8-7-13-24(29(37)38)34-31(40)41-17-23-21-11-3-1-9-19(21)20-10-2-4-12-22(20)23/h1-4,9-12,23-26,28H,5-8,13-18H2,(H,32,36)(H,34,40)(H,37,38)(H2,33,35,39)/t24-,25-,26-,28-/m0/s1
SMILES OC(=O)[C@H](CCCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)NC(=O)OCC3c4ccccc4c5ccccc35
IUPAC Name (2S)-6-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
Molecular Weight (g/mol) 594.72
Formula Weight 594.2512
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.