missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-Nepsilon-benzyloxycarbonyl-L-lysine, 98%

Product Code. 15449318
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
15449318 5 g 5g
15459318 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 15449318

Brand: Thermo Scientific Alfa Aesar H62866.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86060-82-4 | C29H30N2O6 | 502.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86060-82-4
Molecular Formula C29H30N2O6
Molecular Weight (g/mol) 502.57
MDL Number MFCD00065662
InChI Key KRULQRVJXQQPQH-CYLJNIGPNA-N
Synonym fmoc-lys z-oh, fmoc-lys cbz-oh, n, a-fmoc-n, a-z-l-lysine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-6-phenylmethoxycarbonylamino hexanoic acid, 2s-6-benzyloxy carbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, lysine derivative 4, pubchem19681, fmoc-l-lys z-oh, fmoc-n-epsilon-cbz-l-lysine, n alpha-fmoc-n epsilon-z-l-lysine
PubChem CID 3013736
SMILES OC(=O)[C@H](CCCCNC(=O)OCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 110°C to 120°C
Quantity 5 g
Beilstein 3578530
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-(phenylmethoxycarbonylamino)hexanoic acid
Formula Weight 502.56
Percent Purity 98%
Chemical Name or Material Nalpha-Fmoc-Nepsilon-benzyloxycarbonyl-L-lysine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.