missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-N^w-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine, 96%

Product Code. 15445545
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15445545

Brand: Thermo Scientific Alfa Aesar H62148.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 200124-22-7 | C24H38N4O7S | 526.649 g/mol

Nalpha-Boc-Nμ-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine is used as a triflating reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Boc-Nµ-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine is used as a triflating reagent.

Solubility
Soluble in water or 1% acetic acid

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 200124-22-7
Molecular Formula C24H38N4O7S
Molecular Weight (g/mol) 526.649
MDL Number MFCD00236821
InChI Key CVFXPOKENLGCID-KRWDZBQOSA-N
Synonym boc-arg pbf-oh, s-2-tert-butoxycarbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, n-boc-n'-2,2,4,6,7-pentamethyldihydrobenzofuran-5-sufonyl-l-arginine, ambotzbaa1067, boc-arg pbf-0h, n,a-boc-n, o-pbf-l-arginine, boc-arg pbf-oh tlc, o-2,2,4,6,7-pentamethyl-dihydrobenzofuran-5-sulfonyl-l-arginine, 2s-2-tert-butoxycarbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, 2s-5-e-amino 2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-sulfonamido methylidene amino-2-tert-butoxycarbonyl amino pentanoic acid
PubChem CID 11341472
IUPAC Name (2S)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N)C)CC(O2)(C)C

Specifications

Quantity 5 g
Solubility Information Soluble in water or 1% acetic acid
Formula Weight 526.65
Percent Purity 96%
Chemical Name or Material Nalpha-Boc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.