missing translation for 'onlineSavingsMsg'
Learn More

NAlpha-Boc-L-arginine Hydrochloride, TRC

Product Code. 30343604
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30343604 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30343604 Supplier Toronto Research Chemicals Supplier No. TRCB6213255G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nalpha-Boc-L-arginine Hydrochloride
CAS 35897-34-8
Molecular Formula C11H22N4O4 . HCl
Quantity 5 g
Synonym N2-[(1,1-Dimethylethoxy)carbonyl]-L-arginine Hydrochloride, N2-Carboxy-L-arginine N2-tert-Butyl Ester Hydrochloride, (S)-2-[(tert-Butoxycarbonyl)amino]-5-guanidinopentanoic Acid Hydrochloride, N-tert-Butoxycarbonyl-L-arginine Hydrochloride, N-alpha-Butyloxycarbonyl-L-arginine Hydrochloride, N2-tert-Butoxycarbonyl-L-arginine Hydrochloride, NSC 334363
InChI Formula InChI=1S/C11H22N4O4.ClH/c1-11(2,3)19-10(18)15-7(8(16)17)5-4-6-14-9(12)13;/h7H,4-6H2,1-3H3,(H,15,18)(H,16,17)(H4,12,13,14);1H/t7-;/m0./s1
SMILES Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(=N)N)C(=O)O
IUPAC Name (2S)-5-carbamimidamido-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrochloride
Molecular Weight (g/mol) 274.32
Formula Weight 310.1408
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.