missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-D-lysine, 98%

Product Code. 15439408
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15439408 1 g 1g
15459408 250 mg 250mg
15449408 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15439408 Supplier Thermo Scientific Alfa Aesar Supplier No. H62966.03
  • ONLINE EXCLUSIVE PRICE
    £44.00
    £39.65 / 1g
    Save £4.35
    9% Off
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 106719-44-2 | C11H22N2O4 | 246.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106719-44-2
Molecular Formula C11H22N2O4
Molecular Weight (g/mol) 246.31
MDL Number MFCD00076956
InChI Key DQUHYEDEGRNAFO-SVGMAFHSNA-N
Synonym boc-d-lys-oh, boc-l-lys-oh, n2-boc-d-lysine, nalpha-tert-butoxycarbonyl-d-lysine, nalpha-boc-d-lysine, n-, a-boc-d-lysine, boc-d-lysine, d-lysine, n2-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl-d-lysine, n-alpha-t-butoxycarbonyl-d-lysine
PubChem CID 7018770
IUPAC Name (2R)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CCCCN)C(O)=O

Specifications

Melting Point 196°C to 200°C
Quantity 1 g
Optical Rotation −22° (c=2 in Methanol)
Formula Weight 246.3
Percent Purity 98%
Chemical Name or Material Nalpha-Boc-D-lysine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.