missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAlpha-Boc-D-histidine, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Nalpha-Boc-D-histidine |
| CAS | 50654-94-9 |
| Molecular Formula | C11H17N3O4 |
| Quantity | 100 mg |
| Synonym | N-[(1,1-Dimethylethoxy)carbonyl]-D-histidine |
| InChI Formula | InChI=1S/C11H17N3O4/c1-11(2,3)18-10(17)14-8(9(15)16)4-7-5-12-6-13-7/h5-6,8H,4H2,1-3H3,(H,12,13)(H,14,17)(H,15,16)/t8-/m1/s1 |
| SMILES | O=C(O)[C@H](NC(OC(C)(C)C)=O)CC1=CN=CN1 |
| IUPAC Name | (tert-butoxycarbonyl)-D-histidine |
| Molecular Weight (g/mol) | 255.27 |
| Formula Weight | 255.12 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction