Learn More
Alfa Aesar™ Nalpha-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine dicyclohexylammonium salt, 95%
Brand: Alfa Aesar™ H63222.06
Code : NEW
Additional Details : CAS Number : 16879-90-6 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
Nalpha-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine dicyclohexylammonium salt | |
552.74 | |
Z-Tyr(tBu)-OH.DCHA |
16879-90-6 | |
MFCD00038245 |
Specifications
Nalpha-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine dicyclohexylammonium salt | |
Solid | |
552.74 | |
Z-Tyr(tBu)-OH.DCHA |
16879-90-6 | |
95% | |
MFCD00038245 | |
5g |