missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Benzyloxycarbonyl-O-tert-butyl-D-tyrosine dicyclohexylammonium salt, 98%, Thermo Scientific™

Catalog Number 15418106
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
198828-72-7
C33H48N2O5
552.76
MFCD00065702
FDNJRKLIHBJXIR-LKGHUBQFNA-N
z-d-tyr tbu-oh.dcha, z-d-tyr tbu-oh dcha, dicyclohexylamine r-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoate, z-d-tyr tbu-ohcha, z-d-tyr but-oh dcha, z-d-tyr tbu-oh?cha, n-alpha-benzyloxycarbonyl-o-t-butyl-d-tyrosine dicyclohexylamine, 2r-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid; dicha, n-benzyloxy carbonyl-o-tert-butyl-d-tyrosine-n-cyclohexylcyclohexanamine 1/1, n^a-benzyloxycarbonyl-o-tert-butyl-d-tyrosine dicyclohexylammonium salt
53229926
C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC1=CC=C(C[C@@H](NC(=O)OCC2=CC=CC=C2)C(O)=O)C=C1
This item is not returnable. View return policy
198828-72-7
C33H48N2O5
552.76
MFCD00065702
FDNJRKLIHBJXIR-LKGHUBQFNA-N
z-d-tyr tbu-oh.dcha, z-d-tyr tbu-oh dcha, dicyclohexylamine r-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoate, z-d-tyr tbu-ohcha, z-d-tyr but-oh dcha, z-d-tyr tbu-oh?cha, n-alpha-benzyloxycarbonyl-o-t-butyl-d-tyrosine dicyclohexylamine, 2r-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid; dicha, n-benzyloxy carbonyl-o-tert-butyl-d-tyrosine-n-cyclohexylcyclohexanamine 1/1, n^a-benzyloxycarbonyl-o-tert-butyl-d-tyrosine dicyclohexylammonium salt
53229926
C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC1=CC=C(C[C@@H](NC(=O)OCC2=CC=CC=C2)C(O)=O)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}-3-[4-(tert-butoxy)phenyl]propanoic acid; N-cyclohexylcyclohexanamine
Formula Weight 552.74
Percent Purity 98%
Quantity 1 g
Physical Form Solid
Chemical Name or Material Nalpha-Benzyloxycarbonyl-O-tert-butyl-D-tyrosine dicyclohexylammonium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'