missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Benzyloxycarbonyl-L-ornithine, 98%

Product Code. 15429518
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15429518

Brand: Thermo Scientific Alfa Aesar H63052.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2640-58-6 | C13H18N2O4 | 266.297 g/mol

Nalpha-Benzyloxycarbonyl-L-ornithine is widely used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Benzyloxycarbonyl-L-ornithine is widely used as pharmaceutical intermediate.

Solubility
Soluble in 0.1N NaOG (1 mg/ml-clear, colorless solution). Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2640-58-6
Molecular Formula C13H18N2O4
Molecular Weight (g/mol) 266.297
MDL Number MFCD00038200
InChI Key ZYGRWJVRLNJIMR-NSHDSACASA-N
Synonym z-orn-oh, cbz-l-ornithine, cbz-orn-oh, l-ornithine, n2-phenylmethoxy carbonyl, 2s-5-amino-2-benzyloxy carbonyl amino pentanoic acid, s-5-amino-2-benzyloxycarbonylamino pentanoic acid, s-5-amino-2-benzyloxy carbonyl amino pentanoic acid, pubchem18987, carbobenzyloxy-l-ornithine, na-carbobenzoxy-l-ornithine
PubChem CID 7010490
IUPAC Name (2S)-5-amino-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CCCN)C(=O)O

Specifications

Color White
Solubility Information Soluble in 0.1N NaOG (1mg/ml-clear,colorless solution). Slightly soluble in water.
Quantity 1 g
Formula Weight 266.29
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nalpha-Benzyloxycarbonyl-L-ornithine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.