missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Benzyloxycarbonyl-D-asparagine, 95%, Thermo Scientific™

Product Code. 15428978
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428978 25 g 25g
15408978 1 g 1g
15418978 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15428978 Supplier Thermo Scientific Alfa Aesar Supplier No. H62459.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4474-86-6
Molecular Formula C12H14N2O5
Molecular Weight (g/mol) 266.25
MDL Number MFCD00065696
InChI Key FUCKRCGERFLLHP-UHFFFAOYNA-N
Synonym n-cbz-d-asparagine, z-d-asn-oh, nalpha-cbz-d-asparagine, z-d-asparagine, cbz d-asparagine, cbz-d-asn-oh, n-carbobenzoxy-d-asparagine, n-cbz-d-asn-oh, nalpha-carbobenzoxy-d-asparagine, 2r-2-benzyloxy carbonyl amino-3-carbamoylpropanoic acid
PubChem CID 1712147
SMILES NC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 165°C
Quantity 25 g
Optical Rotation −7° (c=1.6 in Acetic Acid)
IUPAC Name (2R)-4-amino-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid
Formula Weight 266.25
Percent Purity 95%
Chemical Name or Material Nalpha-Benzyloxycarbonyl-D-asparagine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.