missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Benzoyl-L-arginine ethyl ester hydrochloride, 99+%

Product Code. 10259693 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass Bottle
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10259693 50 g Glass Bottle 50g
10070710 2.5 g Glass Bottle 2.5g
10102230 10 g Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10259693 Supplier Thermo Scientific Acros Supplier No. 105700500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C15H22N4O3·HCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C15H22N4O3·HCl
MDL Number MFCD00012579
Synonym BAEE

Specifications

Melting Point 129°C to 136°C
CAS Min % 99.0
CAS Max % 100.0
Color White to Yellow
Individual Impurity 99.0% min. (TLC)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula H2NC(=NH)NH(CH2)3CH(NHCOC6H5)CO2C2H5·HCl
Beilstein 09, III, 1232
Specific Rotation −15.0° to −19.0° (24°C, 589nm) (c=2, water)
Specific Rotation Condition - 11.50 (24.00°C c=2,H2O)
Quantity 50 g
Formula Weight 342.83
Percent Purity ≥99.0%
Physical Form Powder, Crystals or Granules
Chemical Name or Material Nα-Benzoyl-L-arginine ethyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.