missing translation for 'onlineSavingsMsg'
Learn More

NAlpha-(2,4-Dinitro-5-fluorophenyl)-D-alaninamide, TRC

Product Code. 30387297
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387297 50 mg
30387597 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387297 Supplier Toronto Research Chemicals Supplier No. TRCD47980550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132055-99-3
Molecular Formula C9H9FN4O5
Molecular Weight (g/mol) 272.19
Synonym (2R)-2-[(5-Fluoro-2,4-dinitrophenyl)amino]propanamide, (R)-2-[(5-Fluoro-2,4-dinitrophenyl)amino]propanamide
SMILES C[C@@H](Nc1cc(F)c(cc1[N+](=O)[O-])[N+](=O)[O-])C(=O)N

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H9FN4O5/c1-4(9(11)15)12-6-2-5(10)7(13(16)17)3-8(6)14(18)19/h2-4,12H,1H3,(H2,11,15)/t4-/m1/s1
Formula Weight 272.0557
Chemical Name or Material Nalpha-(2,4-Dinitro-5-fluorophenyl)-D-alaninamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.