missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-1-(4,4-Dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Nepsilon-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals

Product Code. 15400107
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15400107

Brand: Thermo Scientific Alfa Aesar H66446.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156648-40-7
Molecular Formula C31H36N2O6
Molecular Weight (g/mol) 532.64
MDL Number MFCD00467662
InChI Key CNDRLQTWKWIHEP-KSYWNVGFNA-N
Synonym dde-lys fmoc-oh, dde-l-lys fmoc-oh, dde-lysine fmoc-oh, s-6-9h-fluoren-9-yl methoxy carbonyl amino-2-1-4,4-dimethyl-2,6-dioxocyclohexylidene ethyl amino hexanoic acid, n-a-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n-e-fmoc-l-lysine, n-alpha-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n-epsilon-fmoc-l-lysine, 2s-2-1-4,4-dimethyl-2,6-dioxocyclohexylidene ethyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzdaa1015, n-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n'-fmoc-l-lysine, n-, a-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n-, a-fmoc-l-lysine
PubChem CID 51340473
IUPAC Name (2S)-2-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)ethylamino]-6-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SMILES CC(N[C@@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)=C1C(=O)CC(C)(C)CC1=O

Specifications

Color White
Quantity 1 g
Formula Weight 532.64
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nalpha-1-(4,4-Dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Nepsilon-Fmoc-L-lysine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.