missing translation for 'onlineSavingsMsg'
Learn More

NALP8 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18429707 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429707 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429707 Supplier Novus Biologicals Supplier No. NBP281969AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NALP8 Polyclonal antibody specifically detects NALP8 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NALP8
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias CLR19.2, NACHT, Leucine Rich Repeat And PYD Containing 8, NACHT, LRR And PYD Containing Protein 8, NACHT, LRR And PYD Domains-Containing Protein 8, NALP-8, NLR Family, Pyrin Domain Containing 8, NLRP8, NOD16, Nucleotide-Binding Oligomerization Domain Protein 16, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 8, PAN4, PYRIN And NACHT-Containing Protein 4
Host Species Rabbit
Immunogen NALP8 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human NALP8. The immunogen is located within amino acids 130 - 180 of NALP8. Amino Acid Squence: LEEGESGKIRRYKNSV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 126205
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.