missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NALP7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12829172
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12829172 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12829172 Supplier Invitrogen™ Supplier No. PA521023

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human brain tissue lysate. PA5-21023 can be used with blocking peptide PEP-1137.

NOD proteins include the apoptosis regulator APAF1 (apoptotic protease activating factor 1) and mammalian NOD-LRR proteins. NALP7, also known as PYPAF3, a member of the related PYRIN-containing APAF1-like proteins (PYPAFs/NALPs) is thought to play a crucial role in cell proliferation. NALP7 has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. It is expressed in numerous tissues including uterus and ovary, with low levels in heart and brain. NALP7 inhibits caspase-1-dependent interleukin-1-b secretion and is a feedback regulator of interleukin-1-b secretion. Defects in the NALP7 gene are known to cause the formation of a hydatidiform mole (HYDM) and reduce the growth of carcinoma cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NALP7
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NLRP7
Gene Accession No. Q8WX94
Gene Alias CLR19.4; HYDM; NACHT, leucine rich repeat and PYD containing 7; NACHT, LRR and PYD containing protein 7; NACHT, LRR and PYD domains-containing protein 7; NALP7; NLR family pyrin domain containing 7; NLR family, pyrin domain containing 7; NLRP7; NOD12; Nucleotide-binding oligomerization domain protein 12; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 7; PAN7; PYPAF3; PYRIN-containing APAF1-like protein 3
Gene Symbols NLRP7
Host Species Rabbit
Immunogen An 18 amino acid peptide near the amino terminus of human NALP7.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199713
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.