missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NALP6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£185.00 - £330.00
Specifications
| Antigen | NALP6 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 29928676), Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18106022
|
Novus Biologicals
NBP2-31372SS |
0.025 mg |
£185.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18788223
|
Novus Biologicals
NBP2-31372 |
£330.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
NALP6 Polyclonal specifically detects NALP6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| NALP6 | |
| Western Blot 1-2 ug/ml, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 29928676), Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| P59044 | |
| 171389 | |
| Partial recombinant protein made to an N-terminal portion of human NALP6 (between amino acids 10-200) [UniProt P59044]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Immunology, Inflammation | |
| PBS with 0.05% Sodium Azide | |
| Angiotensin II/Vasopressin Receptor, AVR, CLR11.4, NACHT, Leucine Rich Repeat And PYD Containing 6, NACHT, LRR And PYD Containing Protein 6, NACHT, LRR And PYD Domains-Containing Protein 6, NAVR, NAVR/AVR, NLR Family, Pyrin Domain Containing 6, NLRP6 Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 6, PAN3, PYPAF5, PYRIN-Containing APAF1-Like Protein 5 | |
| NLRP6 | |
| IgG | |
| Protein A purified | |
| Human. Immunogen sequence similarity with other species: Rat (72%), Mouse (70%) . |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title