missing translation for 'onlineSavingsMsg'
Learn More

NALP4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18316243
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18316243 25 μg 25µL
18383076 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18316243 Supplier Bio-Techne Supplier No. NBP31719125UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NALP4 Polyclonal antibody specifically detects NALP4 in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NALP4
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias Cancer/testis antigen 58, CLR19.5, CT58PYRIN and NACHT-containing protein 2, FLJ32126, NACHT, leucine rich repeat and PYD containing 4, NACHT, LRR and PYD domains-containing protein 4, NALP4NACHT, LRR and PYD containing protein 4, NLR family, pyrin domain containing 4, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 4, PAN2Ribonuclease inhibitor 2, PYPAF4PAAD and NACHT-containing protein 2, RNH2PYRIN-containing APAF1-like protein 4
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: FTLTKLSRDDIRSLCDALNYPAGNVKELALVNCHLSPIDCEVLAGLLTNNKKLTYLNVSCNQLDTGVPLLCEALCSPDTVLVYLMLAFCHLS
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147945
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.