missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NALP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572910
This item is not returnable.
View return policy
Description
NALP4 Polyclonal Antibody for Western Blot, IHC (P), IP
NALP proteins include the apoptosis regulator APAF1 (apoptotic protease activating factor 1) and mammalian NOD-LRR proteins and are thought to be involved in inflammation and reproduction. NALP4, also known as PYRIN-containing APAF1-like protein 4, has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. In transfected 293 cells, NALP4 suppressed NF-kappa-B induction by TNF-a and IL-1-b, suggesting NALP4 operates at a point of convergence in these two signaling pathways. NALP4 also suppressed NF-kappa-B induction resulting from overexpression of several adapter proteins and protein kinases involved in these pathways, including TRAF2, TRAF6, RIP, and IRAK2, as well as the IKK-alpha and IKK-beta, suggesting that NALP4 is critical in modulating NF-kappa-B activity.
Specifications
| NALP4 | |
| Polyclonal | |
| Unconjugated | |
| NLRP4 | |
| 4930406H16Rik; cancer/testis antigen 58; CLR19.5; CT58; E330021B02; E330028A19Rik; NACHT, leucine rich repeat and PYD containing 4; NACHT, leucine rich repeat and PYD containing 4A; NACHT, leucine rich repeat and PYD containing 4E; NACHT, LRR and PYD containing protein 4; NACHT, LRR and PYD domains-containing protein 4; NACHT, LRR and PYD domains-containing protein 4A; NACHT, LRR and PYD domains-containing protein 4E; NACHT/LRR/pyrin domain-containing protein 4; NALP, epsilon; NALP4; Nalp4a; Nalp4e; NALP-epsilon; Nalp-eta; Nalp-ita; NLR family pyrin domain containing 4; NLR family, pyrin domain containing 4; NLR family, pyrin domain containing 4A; NLR family, pyrin domain containing 4E; Nlrp4; Nlrp4a; Nlrp4e; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 4; PAAD and NACHT-containing protein 2; PAN2; PYPAF4; PYRIN and NACHT-containing protein 2; PYRIN-containing APAF1-like protein 4; RGD1561918; RGD1563529; Ribonuclease inhibitor 2; RNH2 | |
| Rabbit | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Conc. Not Determined | |
| Whole serum with 0.02% sodium azide | |
| Q96MN2 | |
| NLRP4 | |
| A synthetic peptide corresponding to amino acids 139-157 of human NALP4/PAN2/PYPAF4. | |
| RUO | |
| 147945 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction