missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NALP12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572911
This item is not returnable.
View return policy
Description
NALP12 Polyclonal Antibody for Western Blot, IHC (F), IHC (P), IP
NALP proteins are cytoplasmic proteins that form a subfamily within the larger CATERPILLER family and are thought to play a crucial role in cell proliferation and reproduction. Like all other NALP family members, NALP12, also known as Monarch-1, has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. NALP12 is thought to act as an attenuating factor of inflammation by suppressing inflammatory responses such as NF-kappa-B activation by TLR-signaling molecules MyD88, IRAK-1, TRAF6 and RIPK1 in activated monocytes. Recent evidence suggests that mutations in NALP12 result in hereditary periodic fever syndromes.
Specifications
| NALP12 | |
| Polyclonal | |
| Unconjugated | |
| NLRP12 | |
| CLR19.3; FCAS2; monarch 1; monarch-1; NACHT, leucine rich repeat and PYD containing 12; NACHT, LRR and PYD containing protein 12; NACHT, LRR and PYD domains-containing protein 12; NALP12; NLR family pyrin domain containing 12; NLR family, pyrin domain containing 12; Nlrp12; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 12; PAN6; PYPAF7; PYRIN-containing APAF1-like protein 7; regulated by nitric oxide; RNO; RNO2 | |
| Rabbit | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Conc. Not Determined | |
| Whole serum with 0.05% sodium azide | |
| P59046 | |
| NLRP12 | |
| A synthetic peptide corresponding to amino acids 190-208 of human NALP12/PYPAF7/Monarch-1. | |
| RUO | |
| 91662 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction