missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NALP12 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76293H
This item is not returnable.
View return policy
Description
NALP12 Polyclonal antibody specifically detects NALP12 in Human, Mouse, Rat, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| NALP12 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Virus | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| FCAS2, Monarch-1, NACHT, leucine rich repeat and PYD containing 12, NACHT, LRR and PYD domains-containing protein 12, NALP12LRR and PYD containing protein 12, NLR family, pyrin domain containing 12, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 12, PYPAF7Monarch1, PYRIN-containing APAF1-like protein 7, Regulated by nitric oxide, RNO2 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human NALP12. The immunogen is located within amino acids 110 - 160 of NALP12. Amino Acid Squence: TPRKDPQETYRDYVRRKF | |
| 0.1 mL | |
| Apoptosis | |
| 91662 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction