missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NALP11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561235
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: HLGNDGVAKL LESLISPDCV LKVVGLPLTG LNTQTQQLLM TVKERKPSLI FLSETWSLKE GREIGVTPAS QPGSIIPNSN LDYMFFKFPR MSAAMRTSNT ASR Highest antigen sequence identity to the following orthologs: Mouse - 23%, Rat - 23%.
NALP11 is a 1033 amino acid containing cytoplasmic protein belonging to the NLRP family with a DAPIN domain, six LRR (leucine-rich) repeats and a NACHT domain. Reports suggest that it is a short NALP involved in inflammation potential. NALP proteinsare implicated in the activation of proinflammatory caspases (e.g., CASP1) via their involvement in multiprotein complexes called inflammasomes. The DAPIN domain is necessary and sufficient for suppression of NF-kappa B activation induced by TNF and for inducing IL-1B secretion in collaboration with Caspase-1. It is involved in interaction with PYCARD.
Spezifikation
| NALP11 | |
| Polyclonal | |
| Unconjugated | |
| NLRP11 | |
| CLR19.6; NACHT, leucine rich repeat and PYD containing 11; NACHT, LRR and PYD domains-containing protein 11; NALP11; NLR family pyrin domain containing 11; NLR family, pyrin domain containing 11; NLRP11; NOD17; Nucleotide-binding oligomerization domain protein 17; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 11; PAAD- and NACHT-containing protein 10B; PAAD-and NACHT domain-containing protein 10; PAN10; PYPAF6; PYPAF7; PYRIN-containing APAF1-like protein 6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 204801 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P59045 | |
| NLRP11 | |
| Recombinant protein corresponding to Human NALP11. Recombinant protein control fragment (Product #RP-99200). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction