Learn More
NALP1 Monoclonal Antibody (Nalpy1-4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA125842
Description
MA1-25842 detects NALP1 from human samples. MA1-25842 has been successfully used in immunocytochemistry, immunohistochemistry (frozen), immunohistochemistry (paraffin), immunoprecipitation, immunofluorescence and Western blot procedures. The MA1-25842 immunogen is recombit, full length human NALP1 (pyrin domain).
This gene encodes a member of the Ced-4 family of apoptosis proteins. Ced-family members contain a caspase recruitment domain (CARD) and are known to be key mediators of programmed cell death. The encoded protein contains a distinct N-terminal pyrin-like motif, which is possibly involved in protein-protein interactions. This protein interacts strongly with caspase 2 and weakly with caspase 9. Overexpression of this gene was demonstrated to induce apoptosis in cells. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity of some variants has not been determined.
Specifications
| NALP1 | |
| Monoclonal | |
| 1 mg/mL | |
| NLRP1 | |
| CARD7, CLR17.1, DEFCAP, DEFCAP-L/S, DKFZp586O1822, KIAA0926, NAC, NALP1, PP1044, SLEV1, VAMAS1 | |
| Mouse | |
| Igg Fraction | |
| RUO | |
| 22861 | |
| 4° C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Nalpy1-4 | |
| Unconjugated | |
| Q9C000 | |
| NLRP1 | |
| Recombit, full length human NALP1 (pyrin domain). | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.