missing translation for 'onlineSavingsMsg'
Learn More

Nalidixic acid, MedChemExpress

Product Code. 16460908
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460908 5 g 5g
16438478 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16460908 Supplier Medchem Express Supplier No. HYB0398
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nalidixic acid, a quinolone antibiotic, is effective against both gram-positive and gram-negative bacteria. Nalidixic acid acts in a bacteriostatic manner in lower concentrations and is bactericidal in higher concentrations. Nalidixic acid inhibits a subunit of DNA gyrase and topoisomerase IV and reversibly blocks DNA replication in susceptible bacteria.

Nalidixic acid, a quinolone antibiotic, is effective against both gram-positive and gram-negative bacteria. Nalidixic acid acts in a bacteriostatic manner in lower concentrations and is bactericidal in higher concentrations. Nalidixic acid inhibits a subunit of DNA gyrase and topoisomerase IV and reversibly blocks DNA replication in susceptible bacteria.

Specifications

Chemical Name or Material Nalidixic acid
Physical Form Powder
CAS 389-08-2
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C12H12N2O3
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : 6 mg/mL (25.84 mM; Need ultrasonic) ∣H2O : 5 mg/mL (21.53 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(C1=CN(CC)C2=C(C=CC(C)=N2)C1=O)O
Molecular Weight (g/mol) 232.24
Formula Weight 232.24
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.