missing translation for 'onlineSavingsMsg'
Learn More

Nalidixic Acid, LoGiCal

Product Code. 30459862
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459862 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459862 Supplier LoGiCal Supplier No. LGCFOR3144.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nalidixic acid
CAS 389-08-2
Analyte or Component Names Nalidixic acid
Shipping Condition Room Temperature
Molecular Formula C12 H12 N2 O3
Certifications/Compliance ISO 9001
Synonym Nalidixic acid, 1-ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid, 1,8-Naphthyridine-3-carboxylic acid, 1-ethyl-1,4-dihydro-7-methyl-4-oxo-1,4-Dihydro-1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid, 1-Ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid, 1-Ethyl-7-methyl-1,8-naphthyridin-4-one-3-carboxylic acid, 1-Ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid, 3-Carboxy-1-ethyl-7-methyl-1,8-naphthyridin-4-one, Betaxina, Cybis, Dixiben, Eucistin, Innoxalomn, NSC 82174, Nacid, Nalidic acid, Nalidicron, Nalidixan, Nalidixic acid, Nalidixin, Nalidixinic acid, Nalitucsan, Nalix, Nalurin, Narigix, Naxuril, NegGram, Nelidix, Nevigramon, Nicelate, Nogram, Poleon, Specifin, Uralgin, Uriben, Uriclar, Urisal, Urodixin, Uroman, Uroneg, Uropan, Win 18320, Wintomylon
InChI Formula InChI=1S/C12H12N2O3/c1-3-14-6-9(12(16)17)10(15)8-5-4-7(2)13-11(8)14/h4-6H,3H2,1-2H3,(H,16,17)
Solution Type Neat
SMILES CCN1C=C(C(=O)O)C(=O)c2ccc(C)nc12
IUPAC Name 1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 232.24
Quantity 10 mg
Formula Weight 232.0848 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.