missing translation for 'onlineSavingsMsg'
Learn More

Nalidixic Acid, Fisher BioReagents

Product Code. 10777024
Change view
Click to view available options
Quantity:
25 g
Packaging:
Poly Bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10777024 25 g Poly Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10777024 Supplier Fisher BioReagents Supplier No. BP90825

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Nalidixic acid is an antibiotic that inhibits growth and reproduction primarily in gram-negative bacteria, with some effectiveness against gram-positive bacteria
  • Blocks DNA replication in susceptible bacteria by interfering with DNA gyrase activity
  • Can be used as a selective agent as some bacteria are resistant to nalixidic acid
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Specifications

CAS 389-08-2
Melting Point 225°C
Color White
Loss on Drying 0.5% max.
Melting Range 225°C to 231°C
Assay Percent Range ≥98 %
Packaging Poly Bottle
Molecular Formula C12H12N2O3
Quantity 25 g
Merck Index 15, 6444
Synonym nalidixic acid, nalidixin, nevigramon, nalidixate, uronidix, innoxalon, nalidixan, nalitucsan, sicmylon, unaserus
InChI Key MHWLWQUZZRMNGJ-UHFFFAOYSA-N
SMILES CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)O
IUPAC Name 1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid
Molecular Weight (g/mol) 232.239
PubChem CID 4421
ChEBI CHEBI:100147
Formula Weight 232.24
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Nalidixic Acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nalidixic acid
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.