missing translation for 'onlineSavingsMsg'
Learn More

Melford Nalidixic Acid

Product Code. 30018582
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018582 5 g 5g
30018581 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018582 Supplier Melford Supplier No. N420005.0
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nalidixic Acid Sodium Salt is a chemical compound that is used for its antimicrobial properties and has various applications in microbiology, and pharmaceutical research.

Nalidixic Acid Sodium Salt is a chemical compound that is used for its antimicrobial properties and has various applications in microbiology, and pharmaceutical research. Nalidixic Acid Sodium Salt is employed as a reference standard and selective agent in microbiological culture media. It inhibits the growth of susceptible bacteria, making it a valuable tool for studying bacterial susceptibility and resistance patterns.

Researchers use it to assess the effectiveness of antibiotics against different bacterial strains, contributing to our understanding of microbial biology and antibiotic resistance.

In pharmaceutical research, Nalidixic Acid Sodium Salt can be used as a reference standard for antibiotic testing and formulation. It supports the development and optimization of antibiotics and strategies to combat antibiotic-resistant bacteria.

Specifications

Chemical Name or Material Nalidixic Acid
CAS 389-08-2
Quantity 5 g
Molecular Formula C12H12N2O3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.