missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Wash Bottles
GREENER_CHOICE

Product Code. 10643062
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Closure Size:
53 mm
63 mm
Unit Size:
Pack of 2
Pack of 4

Product Code. 10643062

Brand: Thermo Scientific™ 24071000

Only null left
Add to basket
Edge
Add to basket

Pour without sputter with these wide-mouth LDPE wash bottles which feature through-the-top closure and stem molded in one piece.

High quality squeeze bottles with wide mouth for easy filling. Thermo Scientific™ Nalgene™ Wide Mouth LDPE Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.

  • Leakproof† bottle comes with polypropylene copolymer draw tube and polypropylene screw closure.
  • Tip can be cut back for increased flow.
  • Tight fit of draw tube and closure prevents separation when squeezing bottle.
TRUSTED_SUSTAINABILITY

Specifications

Capacity 32 oz., 1000 mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Translucent
Description Polypropylene copolymer draw tube
Material LDPE
No. per Case 12
No. per Pack 2
Closure Included Yes
Graduated No
Mouth Wide
Shape Cylindrical
Height (Metric) 246 mm
Product Type Wash Bottle
Height (English) 9.7 in.
Closure Material Polypropylene
Closure Size 63 mm
Diameter (Metric) 92.2 mm
Closure Type Screw Cap
Through the Closure Dispensing Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.