Learn More
Thermo Scientific™ Nalgene™ Tubing Y-Type Connectors
Maximize liquid flow with non-tapered hose barbed Y-Type Connectors, molded of polypropylene resin, free of animal derived components.
Brand:  Thermo Scientific™ 6152-0312
Code : 55
Additional Details : Weight : 0.02880kg
 View more versions of this product
Product Code. 10020601
  • £70.50 / Pack of 12
In Stock
to see stock.
Add to basket
Description
Description
Maximize liquid flow with non-tapered hose barbed Thermo Scientific™ Nalgene™ Tubing Y-Type Connectors, molded of polypropylene resin, free of animal derived components. Products are double-bagged to maintain cleanliness.
  • Polypropylene construction
  • Autoclavable
Specifications
Specifications
Tubing Connector
Autoclavable
7.93 mm
5/16 in. I.D.
Y-Type Connector
5/16 in.
Polypropylene
12/Pk., 72/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.