Learn More
Thermo Scientific™ Nalgene™ Torque Wrench Fittings for Polypropylene Closure
Product Code. 10314293
Change view
Click to view available options
Closure Size:
11 mm (High Profile MPV)
11 mm (Low Profile MPV)
13 to 415 mm
20 to 415 mm
24 to 415 mm
28 to 415 mm
38 to 415 mm
38 to 430 mm
43 to 415 mm
48 to 415 mm
53 to 415 mm
53B
63 to 415 mm
83B
Unit Size:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size unitSize
10314293 38 to 430 mm Each
10680904 11 mm (Low Profile MPV) Each
10304533 11 mm (High Profile MPV) Each
10542035 13 to 415 mm Each
10777093 20 to 415 mm Each
10436853 24 to 415 mm Each
10542995 28 to 415 mm Each
10314723 38 to 415 mm Each
10707103 43 to 415 mm Each
10651674 48 to 415 mm Each
10777273 53 to 415 mm Each
10232863 63 to 415 mm Each
10543955 53B Each
10282283 83B Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 10314293 Supplier Thermo Scientific™ Supplier No. 21950438
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Achieve accurate torque readings when applying closures onto bottles with specific torque wrench fittings for polypropylene closures.
Achieve accurate torque readings when applying closures onto bottles with Thermo Scientific™ Nalgene™ Torque Wrench Fittings for Polypropylene Closures. Proper torquing of Nalgene closures is critical to leakproof† sealing.
  • Epoxy
  • Specifically designed for Nalgene polypropylene closures
Compatible with:
Thermo Scientific Nalgene Torque Wrenches (2195-6169 and -6177) † The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fittings
Closure Size 38 to 430 mm
Drive Size 1/4 in.
For Use With (Equipment) Nalgene Torque Wrenches (Cat. Nos. 2195-6169 and -6177)
Material Epoxy
No. per Pack 1 Ea.