missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene Torque Fitting Tool for Nalgene Bottle and Closure Combinations

Product Code. 32188523
Change view
Click to view available options
For Use With (Equipment):
13 mm Closures
20 mm Closures
20 mm Serum Vial Closures, HDPE
24 mm Closures
28 mm Closures
38 mm Biotainer Closures
38-415 Closures
38-430 Closures
43 mm Closures
48 mm Biotainer Closures
48 mm Closures
53 mm Closures
53B Closures
63 mm Closures
83B Closures
Unit Size:
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
32188523 48 mm Biotainer Closures Each
32188525 13 mm Closures Each
32188520 20 mm Closures Each
32188527 24 mm Closures Each
32188530 28 mm Closures Each
32188524 38-415 Closures Each
32188517 43 mm Closures Each
32188521 48 mm Closures Each
32188529 53 mm Closures Each
32188526 63 mm Closures Each
32188522 53B Closures Each
32188519 83B Closures Each
32188528 38-430 Closures Each
32188518 20 mm Serum Vial Closures, HDPE Each
32188531 38 mm Biotainer Closures Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 32188523 Proveedor Thermo Scientific™ N.º de proveedor 31959048
Estimated Shipment: 20-11-2026
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Ensure a guaranteed leak-proof seal with proper torque settings.

The Nalgene Bottle Torque Fittings represent a specialized closure solution engineered to deliver precise torque application and guaranteed leakproof performance across Nalgene's laboratory vessel portfolio. These fittings are uniquely designed for compatibility with various Nalgene bottle and closure configurations, including lab bottles, packaging bottles, media bottles, biotainers, and PETG serum vials.

  • Comprehensive Size Range: Available in multiple sizes engineered to accommodate the majority of standard laboratory bottles and closure configurations
  • Multi-Material Closure Compatibility: Designed for optimal performance with both high-density polyethylene (HDPE) and polypropylene (PP) closure materials
  • Nalgene-Specific Design: Custom-engineered geometry specifically optimized for Nalgene laboratory closure systems
  • Universal Torque Tool Integration: Compatible with industry-standard 1/4 in. and 3/8 in. square drive torque tools for precise torque application
  • Manual and Automated Operation: Supports both manual torque application and automated torque systems for laboratory workflow integration
  • Product Specifications and Operating Parameters - Detailed torque settings are available in the documents section of this page.
  • Comprehensive Documentation Provides Essential Operating Procedures - Detailed operational instructions are provided in the accompanying documentation section to ensure proper implementation and use of the bottle and closure system.
TRUSTED_SUSTAINABILITY

Especificaciones

For Use With (Application) Proper torque of closures
For Use With (Equipment) 48 mm Biotainer Closures
Product Type Torque Fitting
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.