missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Syringe Filters, Non-sterile
GREENER_CHOICE

Product Code. 10353291
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
0.8 μm
1.1 μm
Diameter (Metric):
4 mm
13 mm
25 mm
Material (Membrane):
Cellulose Acetate
Glass Fiber
Nylon
PTFE
SFCA
Unit Size:
Pack of 100
Pack of 50
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Diameter (Metric) Material (Membrane) unitSize
10353291 0.2 μm 4 mm Nylon Pack of 100
15362378 0.2 μm 13 mm PTFE Pack of 100
15347006 0.45 μm 13 mm PTFE Pack of 100
15372378 1.1 μm 25 mm Glass Fiber Pack of 50
15352388 0.2 μm 25 mm SFCA Pack of 100
15362388 0.45 μm 25 mm SFCA Pack of 100
15382388 0.8 μm 25 mm Cellulose Acetate Pack of 100
15342398 0.2 μm 25 mm Nylon Pack of 100
15352398 0.45 μm 25 mm Nylon Pack of 100
15362398 0.2 μm 25 mm PTFE Pack of 100
15372398 0.45 μm 25 mm PTFE Pack of 100
10722732 0.45 μm 4 mm Nylon Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 10353291 Supplier Thermo Scientific™ Supplier No. 1760020
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Filter sample volumes of 10 to 100mL with these nonsterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.

Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.

Key Features
  • 10 to 100mL sample size
  • Inlet: Female Luer-Lok™
  • Outlet: Male Luer slip
  • Polypropylene housing with ID color rings
  • Pore size and membrane type printed on unit

Surfactant-free Cellulose Acetate (SFCA)

  • Low protein binding, lower extractables than standard CA
  • Good for cell culture fluids and cellular solutions
  • Polypropylene housings with blue color rings

Cellulose Acetate (CA)

  • Some protein binding and extractables
  • For sterile filtration and clarification of: buffers, aqueous solutions and reagents
  • Polypropylene housings with red color rings

Nylon

  • Naturally hydrophilic, low extractables, some protein binding
  • Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
  • Polypropylene housings with pink color rings
PTFE
  • Naturally hydrophobic
  • Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
  • Polypropylene housings with white color rings

TRUSTED_SUSTAINABILITY

Specifications

Color Code Purple
Material (Membrane) Nylon
Pore Size 0.2 μm
Sterility Non-sterile
For Use With (Application) Sterilizing cryoprotectants such as DMSO and with
Housing Material Polypropylene
Product Line Nalgene
Type Syringe Filter
Volume (Metric) 10 to 100 mL (Sample)
Diameter (Metric) 4 mm
Diameter (Metric) Membrane 4 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.