missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Square Amber PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays
GREENER_CHOICE

Product Code. 10581265
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
Closure Size:
20 mm
24 mm
38-430 mm
Unit Size:
Pack of 200
Pack of 280
Pack of 96
Product Code. Capacity (Metric) Closure Size unitSize
10581265 125 mL 38-430 mm Pack of 96
10049442 30 mL 20 mm Pack of 280
10059442 60 mL 24 mm Pack of 200
3 options
This item is not returnable. View return policy

Product Code. 10581265

Brand: Thermo Scientific™ 3220210125

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Safely store light-sensitive medias, buffers and sera in these square, amber PETG bottles.

Thermo Scientific™ Nalgene Square Amber PETG Media Bottles include HDPE closures. Translucent amber bottles meet USP light-resistant requirements and are ideal for medias, buffers and sera.

  • Heavy-walled, shatter resistant and durable
  • Space-saving square design
  • Molded in graduations
  • Nonsterile, non-pyrogenic, non-cytotoxic
  • Leakproof†, with drip-proof neck finish and molded-in neck ring
  • PETG provides excellent gas barrier, with transparency and reduced permeability to CO2 and O2
  • Amber bottle and closure provide protection for light sensitive reagents

Ordering Information:

Products are shrink-wrapped into trays, then packed in double PE lined cartons for cleanroom applications.

Recommended for:

For storage and shipping of light sensitive liquid media, buffers, and sera

Note:

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 125 mL
Capacity (English) 4.2 oz.
Color Amber
Diameter (English) 2.1 in.
Diameter (English) Inner Neck 1.1 in.
Diameter (Metric) Inner Neck 28 mm
Material PETG
No. per Case 96
No. per Pack 24
Sterility Non-sterile
Certifications/Compliance PETG Media Bottles and HDPE closure materials meet current biological testing standards including USP Class VI.
Closure Included Yes
Graduated Yes
Mouth Wide
Shape Square
Height (Metric) 110 mm
Product Type Media Bottle
Height (English) 4.3 in.
Closure Material HDPE
Closure Size 38-430 mm
Diameter (Metric) 54 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.