missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes
GREENER_CHOICEBIOPHARMA

Product Code. 10790602
Change view
Click to view available options
Pore Size:
0.1 μm
0.2 μm
0.45 μm
0.8 μm
Material (Membrane):
Cellulose Nitrate
Nylon
PES
SFCA
Diameter (Metric) Membrane:
50 mm
75 mm
90 mm
Volume (Metric) Receiver:
50 mL
115 mL
150 mL
250 mL
500 mL
1000 mL
1000mL
Unit Size:
Pack of 12
55 product options available for selection
Product selection table with 55 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Material (Membrane) Diameter (Metric) Membrane Volume (Metric) Receiver unitSize
10790602 0.2 μm Nylon 90 mm 500 mL Pack of 12
10685813 0.2 μm PES 50 mm 50 mL Pack of 12
10418232 0.45 μm PES 50 mm 115 mL Pack of 12
10338763 0.1 μm PES 50 mm 150 mL Pack of 12
10102451 0.2 μm PES 50 mm 115 mL Pack of 12
10370311 0.2 μm PES 50 mm 150 mL Pack of 12
10034550 0.45 μm PES 50 mm 150 mL Pack of 12
10793344 0.1 μm PES 75 mm 250 mL Pack of 12
10421791 0.2 μm PES 50 mm 250 mL Pack of 12
10568632 0.45 μm PES 50 mm 250 mL Pack of 12
10474415 0.1 μm PES 75 mm 500 mL Pack of 12
10229090 0.2 μm PES 75 mm 500 mL Pack of 12
10687541 0.2 μm PES 90 mm 500 mL Pack of 12
10044550 0.45 μm PES 75 mm 500 mL Pack of 12
10462701 0.45 μm PES 90 mm 500 mL Pack of 12
12686366 0.1 μm PES 90 mm 1000 mL Pack of 12
10411791 0.2 μm PES 90 mm 1000 mL Pack of 12
10228340 0.45 μm PES 90 mm 1000 mL Pack of 12
10577322 0.2 μm Cellulose Nitrate 50 mm 115 mL Pack of 12
10589222 0.45 μm Cellulose Nitrate 50 mm 115 mL Pack of 12
10403701 0.2 μm Cellulose Nitrate 50 mm 150 mL Pack of 12
10682741 0.45 μm Cellulose Nitrate 50 mm 150 mL Pack of 12
10684851 0.8 μm Cellulose Nitrate 50 mm 150 mL Pack of 12
10261741 0.2 μm Cellulose Nitrate 50 mm 250 mL Pack of 12
10281881 0.45 μm Cellulose Nitrate 50 mm 250 mL Pack of 12
10792351 0.8 μm Cellulose Nitrate 50 mm 250 mL Pack of 12
10139130 0.2 μm Cellulose Nitrate 75 mm 500 mL Pack of 12
10037720 0.45 μm Cellulose Nitrate 75 mm 500 mL Pack of 12
10720622 0.8 μm Cellulose Nitrate 75 mm 500 mL Pack of 12
10047720 0.2 μm Cellulose Nitrate 75 mm 1000 mL Pack of 12
10693701 0.45 μm Cellulose Nitrate 75 mm 1000 mL Pack of 12
10422941 0.8 μm Cellulose Nitrate 75 mm 1000 mL Pack of 12
10450241 0.2 μm SFCA 50 mm 115 mL Pack of 12
10126070 0.45 μm SFCA 50 mm 115 mL Pack of 12
10454641 0.2 μm SFCA 50 mm 150 mL Pack of 12
10760792 0.45 μm SFCA 50 mm 150 mL Pack of 12
10444831 0.2 μm SFCA 50 mm 250 mL Pack of 12
10579051 0.45 μm SFCA 50 mm 250 mL Pack of 12
10017770 0.2 μm SFCA 75 mm 500 mL Pack of 12
10211561 0.2 μm SFCA 90 mm 500 mL Pack of 12
10199700 0.45 μm SFCA 75 mm 500 mL Pack of 12
10740602 0.45 μm SFCA 90 mm 500 mL Pack of 12
10017820 0.2 μm SFCA 75 mm 500 mL Pack of 12
10382711 0.2 μm SFCA 90 mm 1000 mL Pack of 12
10614861 0.45 μm SFCA 50 mm 500 mL Pack of 12
10405021 0.45 μm SFCA 90 mm 1000 mL Pack of 12
10762732 0.2 μm Nylon 50 mm 150 mL Pack of 12
10159130 0.45 μm Nylon 50 mm 150 mL Pack of 12
10760802 0.2 μm Nylon 50 mm 250 mL Pack of 12
10790412 0.45 μm Nylon 50 mm 250 mL Pack of 12
10578471 0.2 μm Nylon 75 mm 500 mL Pack of 12
10211321 0.45 μm Nylon 75 mm 500 mL Pack of 12
10444261 0.2 μm Nylon 75 mm 1000 mL Pack of 12
10362811 0.2 μm Nylon 90 mm 1000 mL Pack of 12
10169700 0.45 μm Nylon 75 mm 1000mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 55 options available.
55 options
Product Code. 10790602 Supplier Thermo Scientific™ Supplier No. 1630020
Only null left
Add to basket
Edge
Add to basket

Increase filtration efficiency. PES membranes and exclusive Rapid-Flow design provide fast flow rates and high throughput while defending against cell culture contamination.

Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ Disposable Filter Units feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid Flow Filter Units and Bottle Tops are the best choice for the filtration of cell culture media, serum, additives and buffers. Some products on this page are ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions (see Full Specifications in the product table for item participation).

  • Available in PES, CN, SFCA and Nylon membranes. Devices are color coded according to membrane.
  • Devices feature the fastest flow rate and lowest clogging of comparable alternatives
  • 0.1um PES available for mycoplasma protection
  • 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
  • Non-pyrogenic and endotoxin free - Rapid-Flow filter units meet the most rigorous testing standards for any vacuum filtration unit.
  • Available with Fit for Purpose Sterile documentation
  • Some products on this page are ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions (see "Full Specifications" in the product table for item participation)
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Material (Membrane) Nylon
Pore Size 0.2 μm
Sterility Sterile
No. per Case 12
No. per Pack 12
Type Filter Unit
Volume (Metric) Receiver 500 mL
Volume (Metric) Upper 500 mL
Diameter (Metric) Membrane 90 mm
Fit for Purpose Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.