missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
GREENER_CHOICEBIOPHARMA

Product Code. 10526952
Change view
Click to view available options
Volume (Metric):
150 mL
500 mL
1000 mL
Material (Membrane):
PES
SFCA
Pore Size:
0.2 μm
0.45 μm
Fits Bottle Neck Size (Metric):
33mm
45mm
Unit Size:
Pack of 12
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Material (Membrane) Pore Size Fits Bottle Neck Size (Metric) unitSize
10526952 500 mL PES 0.2 μm 33mm Pack of 12
10441411 150 mL PES 0.2 μm 33mm Pack of 12
10598282 150 mL PES 0.2 μm 45mm Pack of 12
10743281 150 mL PES 0.45 μm 33mm Pack of 12
10319590 150 mL PES 0.45 μm 45mm Pack of 12
10300461 500 mL PES 0.2 μm 45mm Pack of 12
10706971 500 mL PES 0.45 μm 33mm Pack of 12
10646421 500 mL PES 0.45 μm 45mm Pack of 12
10491093 1000 mL PES 0.2 μm 33mm Pack of 12
10401103 1000 mL PES 0.2 μm 45mm Pack of 12
10352911 150 mL SFCA 0.2 μm 33mm Pack of 12
10139560 150 mL SFCA 0.2 μm 45mm Pack of 12
10027780 150 mL SFCA 0.45 μm 33mm Pack of 12
10624281 150 mL SFCA 0.45 μm 45mm Pack of 12
10037790 500 mL SFCA 0.2 μm 33mm Pack of 12
10201371 500 mL SFCA 0.2 μm 45mm Pack of 12
10529051 500 mL SFCA 0.45 μm 33mm Pack of 12
10549241 500 mL SFCA 0.45 μm 45mm Pack of 12
10139800 1000 mL SFCA 0.2 μm 33mm Pack of 12
10790592 1000 mL SFCA 0.2 μm 45mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
This item is not returnable. View return policy
CAUTION: Use only sterile bottles designed for use with vacuum. Always use safety shield during vacuum procedures.
Product Code. 10526952 Supplier Thermo Scientific™ Supplier No. 5953320
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
CAUTION: Use only sterile bottles designed for use with vacuum. Always use safety shield during vacuum procedures.

Achieve fast flow rates and high throughput for increased filtration efficiency with Nalgene Rapid-Flow sterile disposable bottle top filters.

Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ single use bottle tops feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid-Flow bottle tops are the best choice for the filtration of cell culture media, serum, additives and buffers.

  • Available in PES and SFCA membranes. Devices are color coded according to membrane.
  • Devices feature the fastest flow rate and lowest clogging of comparable alternatives
  • 0.1um PES available for mycoplasma protection
  • 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
  • Non-pyrogenic and endotoxin free - Rapid-Flow filters meet the most rigorous testing standards for any vacuum filtration unit
  • Available with Fit for Purpose Sterile documentation
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Membrane Filter
Material Polystyrene
Material (Membrane) PES
Pore Size 0.2 μm
Sterility Sterile
Fits Bottle Neck Size (Metric) 33mm
No. per Case 12
No. per Pack 12
Type Bottle Top Filter
Volume (Metric) 500 mL
Diameter (Metric) 75 mm
Diameter (Metric) Membrane 75 mm
Fit for Purpose Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.