missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Autoclavable PPCO Wash Bottles
GREENER_CHOICE

Product Code. 10227830
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Closure Size:
28 mm
38-430 mm
Unit Size:
Pack of 4
Pack of 6

Product Code. 10227830

Brand: Thermo Scientific™ 24051000

Only null left
Add to basket
Add to basket

Reduce the risk of contamination when dispensing sterile water or reagents by using these autoclavable PPCO wash bottles.

Reduce the risk of contamination when dispensing sterile water or reagents by using autoclavable Thermo Scientific™ Nalgene™ PPCO Wash Bottles. Made from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. These squeeze bottles are flexible, contact-clear, and designed to dispense through the top with a closure and stem molded in one piece for reliable service. Bent stem dispensing tube retains its bend after autoclaving but bend angle can be modified by hand while warm if desired. The tight fit of the draw tube and closure prevents separation when squeezing bottle.

  • Polypropylene screw/stem and draw tube
  • Tip can be cut back to provide increased flow
TRUSTED_SUSTAINABILITY

Specifications

Capacity 32 oz., 1000 mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Translucent
Includes Screw/stem and draw tube
Material PPCO
No. per Case 12
No. per Pack 4
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Shape Cylindrical
Height (Metric) 259 mm
Product Type Wash Bottle
Height (English) 10.2 in.
Closure Material Polypropylene
Closure Size 38-430 mm
Diameter (Metric) 91.4 mm
Closure Type Cap
Through the Closure Dispensing Yes
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.