missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene, Wide-Mouth Carboy with Handle

Product Code. 15241799
Click to view available options
Capacity (Metric):
10 L
20 L
Unit Size:
Each
Pack of 4
This item is not returnable. View return policy

Product Code. 15241799

Brand: Thermo Scientific™ 22350050

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For Use With: Store and transport solids, powders and liquids; Autoclavable; Closure Material: Polypropylene; Color: White

Store and transport solids, powders and liquids using Thermo Scientific™ Nalgene™ Wide-Mouth Autoclavable Polypropylene Carboys with Handles. These durable natural polypropylene (PP) carboys feature a convenient wide-mouth opening to promote ease of cleaning and dispensing. The wide-shoulder handles enable easy carrying and pouring, even with gloved hands.

  • Graduated to contain in 1 gal. and 5 L increments for ease of use
  • Wide-mouth opening can accommodate overhead mixers
  • Autoclavable
  • Leakproof† white polypropylene 100 mm screw closures
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Closure Material Polypropylene
Closure Size 100-415 mm
Color White
Description Autoclavable Wide-Mouth Carboy with Handles
Diameter (Metric) Outer 282 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
Graduations Graduated to contain in 5 L/1 gal. increments
Height (Metric) With Closure 483 mm
Includes Handles
Material Polypropylene
Mouth Style Wide
No. per Case 4/Cs.
No. per Pack 1 Ea.
Product Type Carboy with Handle
Sterility Non-sterile
Volume (English) Brim 5.5 gal.
Volume (Metric) Brim 22.5 L
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.