missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Stirring Rod
GREENER_CHOICE

Product Code. 11714743
Change view
Click to view available options
Length (Metric):
248 mm
Unit Size:
Pack of 72
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11714743 248 mm Pack of 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11714743 Supplier Thermo Scientific™ Supplier No. 61690010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Combine functions of a stirring rod and rubber policeman with these polypropylene stirring rods, featuring a flat paddle at one end and a triangular paddle at the other end.

Combine the functions of a stirring rod and rubber policeman with Thermo Scientific™ Nalgene™ Polypropylene Stirring Rods, featuring a flat paddle at one end and a triangular paddle at the other end. These autoclavable stirring rods are chemically compatible with a wide range of lab reagents.

  • Flat paddle, 5/8 x 5/8″ (1.6 x 1.6cm) at one end; triangular paddle at the other end
  • Overall length is 9.75″ (24.8cm), makes it a convenient and conventional length for a wide variety of lab uses
  • Usable over a temperature range of 0°C to +135°C making it suitable for use in heated applications

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Color White
Description Stirring Rod with flat paddle and triangular paddle ends
Length (English) 9¾ in.
Length (Metric) 248 mm
Material Polypropylene
Type Stirring Rod
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.