missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene, Replacement Coupling Inserts, elbow coupling, 1/4 inch

Product Code. 10744053
Click to view available options
Quantity:
Case of 6
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 10744053

Brand: Thermo Scientific™ 21590011

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For Use With: Nalgene Quick Filling/Venting Closures

Promote faster installation using Thermo Scientific™ Nalgene™ Polypropylene Replacement Coupling Inserts for Quick/Filling Venting Closures, which support one-handed connection and disconnection when applied to mating coupling bodies. Male, barbed quick-disconnect replacement coupling inserts for use with Nalgene Quick Filling/Venting Closures.

  • For use with Nalgene Quick Filling/Venting Closures
  • Promotes fewer leakpoints and short assemblies

TRUSTED_SUSTAINABILITY

Specifications

Description 90° Elbow coupling insert
Diameter (English) Inner Tubing 1/4 in.
Diameter (Metric) Inner Tubing 6.35 mm
For Use With (Equipment) Nalgene Quick Filling/Venting Closures
Material Polypropylene
No. per Case 6/Cs.
No. per Pack 6 Pk.
Product Type Insert
Quantity Case of 6
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.