missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polycarbonate, Validation Bottle

Product Code. 10601912
Click to view available options
Capacity (Metric):
30 mL
250 mL
2 L
Unit Size:
Pack of 12
Pack of 30
Pack of 6
This item is not returnable. View return policy

Product Code. 10601912

Brand: Thermo Scientific™ DS21270250

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For Use With: Perform validation testing; Autoclavable; Closure Material: Polypropylene; Color: White (Translucent)

Perform QA lot sample storage of high-valued pharmaceuticals using Thermo Scientific™ Nalgene™ Polycarbonate Validation Bottles. Small-volume bottles are molded of the same materials as larger carboys, but require a lower volume of expensive reagents to perform validation testing.

Autoclavable, transparent polycarbonate construction allows for convenient material compatibility validation.

Includes:
White polypropylene screw closure and TPE gasket

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Capacity (English) 8 oz.
Capacity (Metric) 250 mL
Closure Material Polypropylene
Closure Size 53B
Color White (Translucent)
Diameter (Metric) Inner Neck 45 mm
Diameter (Metric) Outer 74 mm
For Use With (Application) Perform validation testing
Height (Metric) With Closure 135 mm
Material Polycarbonate
Mouth Style Wide
No. per Case 6/Cs.
No. per Pack 6 Pk.
Product Type Validation Bottle
Sterility Non-sterile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.