missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Automatic Burette Clamp

Product Code. 10100212
Click to view available options
Size (Metric) Grip:
0 to 25 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10100212

Brand: Bochem™ 5624

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Hold burettes or thermometers securely with this easy-to-use automatic clamp. Bochem™ Automatic Burette Clamp is chromium-plated for durability.

  • Automatic clamp.
  • Includes shaft.
  • Chromium plated.

Specifications

Quantity 1 Ea.
Type Stand Clamp
Material Chromium-plated
Includes Shaft
For Use With (Equipment) Burette, thermometer 0-25mm in diameter
Diameter (Metric) 8 mm
Length (Metric) Arm 115 mm
Size (Metric) Grip 0 to 25 mm
Jaw Type Automatic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.