missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Serum Vials; with Crimp Finish: Sterile, Shrink-Wrapped Modules
GREENER_CHOICEBIOPHARMA

Product Code. 10292853
Change view
Click to view available options
Volume (Metric):
3 mL
5 mL
10 mL
20 mL
Closure Size:
13 mm
20 mm
Unit Size:
Pack of 1260
Pack of 1932
Pack of 3451
Pack of 612
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Closure Size unitSize
10292853 5 mL 20 mm Pack of 1932
10282473 3 mL 13 mm Pack of 3451
10202863 10 mL 20 mm Pack of 1260
10632824 20 mL 20 mm Pack of 612
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10292853 Supplier Thermo Scientific™ Supplier No. 3420300005
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Protect against leakage, breakage and contamination by using PETG serum vials with crimp finish. Lighter in weight than glass, these vials are made for use with stoppers and caps.

Protect against leakage, breakage, and contamination. Thermo Scientific™ Nalgene PETG Crimp Finish Serum Vials are lighter in weight than glass, and are made for use with stoppers and caps.

Features:

  • PETG
  • Noncytotoxic
  • Nonpyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Sterile to 10-6 SAL

Ordering Information:

Serum vials are packed in trayless shrink-wrapped modules. Stoppers and caps not included.

TRUSTED_SUSTAINABILITY

Specifications

Description Serum Vials
Diameter (Metric) Inner 13 mm
Color Clear
Diameter (Metric) Outer 22 mm
Material PETG
Sterility Sterile
Volume (Metric) 5 mL
No. per Case 1932
No. per Pack 276
Volume (English) 1/6 oz.
Product Line Nalgene
Volume (Metric) Total 5 mL
Closure Type Stoppers and Caps (Not Included)
Quantity 1932 Cases, 276 Pk.
Closure Size 20 mm
For Use With (Equipment) Stopper, Cap
Certifications/Compliance ISO 13485:2016
Height (Metric) 40 mm
Product Type Serum Vial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.